MilliporeSigma
  • Molecular and functional characterisation of two elovl4 elongases involved in the biosynthesis of very long-chain (>C
Molecular and functional characterisation of two elovl4 elongases involved in the biosynthesis of very long-chain (>C
Comparative biochemistry and physiology. Part B, Biochemistry & molecular biology (2017-07-03)
Min Jin, Óscar Monroig, Juan Carlos Navarro, Douglas R Tocher, Qi-Cun Zhou
ABSTRACT
Elongation of very long-chain fatty acid (Elovl) 4 proteins are important fatty acyl elongases that participate in the biosynthesis of long-chain (C
Sigma-Aldrich
Stearidonic acid, ≥99%