MilliporeSigma
  • Physical and Pharmacokinetic Characterizations of trans-Resveratrol (t-Rev) Encapsulated with Self-Assembling Lecithin-based Mixed Polymeric Micelles (saLMPMs).
Physical and Pharmacokinetic Characterizations of trans-Resveratrol (t-Rev) Encapsulated with Self-Assembling Lecithin-based Mixed Polymeric Micelles (saLMPMs).
Scientific reports (2017-09-08)
Tzu-Pin Li, Wan-Ping Wong, Ling-Chun Chen, Chia-Yu Su, Lih-Geeng Chen, Der-Zen Liu, Hsiu-O Ho, Ming-Thau Sheu
ABSTRACT
This study involved physical and pharmacokinetic characterizations of trans-resveratrol (t-Rev)-loaded saLMPMs which attempted to improve t-Rev's pharmacokinetic profiles and bioavailability resolving hurdles limiting its potential health benefits. The optimal formulation consisted of t-Rev, lecithin, and Pluronic
Sigma-Aldrich
Levallorphan tartrate salt, ≥98% (HPLC), powder