MilliporeSigma
  • Liquid coordination complexes of Lewis acidic metal chlorides: Lewis acidity and insights into speciation.
Liquid coordination complexes of Lewis acidic metal chlorides: Lewis acidity and insights into speciation.
Dalton transactions (Cambridge, England : 2003) (2017-08-03)
James M Hogg, Lucy C Brown, Karolina Matuszek, Piotr Latos, Anna Chrobok, Małgorzata Swadźba-Kwaśny
ABSTRACT
Coordination complexes of Lewis acidic metal chlorides AlCl
Sigma-Aldrich
Antimony(III) chloride, ≥99.95% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis