MilliporeSigma
SK4 channels modulate Ca
Molecular oncology (2017-05-31)
Friederike A Steudel, Corinna J Mohr, Benjamin Stegen, Hoang Y Nguyen, Andrea Barnert, Marc Steinle, Sandra Beer-Hammer, Pierre Koch, Wing-Yee Lo, Werner Schroth, Reiner Hoppe, Hiltrud Brauch, Peter Ruth, Stephan M Huber, Robert Lukowski
ABSTRACT
Oncogenic signalling via Ca
Sigma-Aldrich
TRAM-34, ≥98% (HPLC), solid