MilliporeSigma
  • Rutaecarpine alleviates renal ischemia reperfusion injury in rats by suppressing the JNK/p38 MAPK signaling pathway and interfering with the oxidative stress response.
Rutaecarpine alleviates renal ischemia reperfusion injury in rats by suppressing the JNK/p38 MAPK signaling pathway and interfering with the oxidative stress response.
Molecular medicine reports (2017-06-01)
Chunhua Wang, Zongyao Hao, Jun Zhou, Li Zhang, Yexiang Sun, Chaozhao Liang
ABSTRACT
In the present study, the protective effect and the potential underlying mechanism of rutaecarpine (Ru) on renal ischemia reperfusion injury (IRI) in rats were investigated. A renal ischemia reperfusion mouse model was established. Ru at 30, 60 mg/kg administered intraperitoneally prior to reperfusion led to attenuated renal injury. The results demonstrated that Ru treatment significantly reduced the content of serum creatinine, urea nitrogen and neutrophil gelatinase‑associated lipocalin in rats with renal IRI. In addition, Ru treatment improved the degree of renal proximal tubular necrosis, decreased the content of inflammatory cytokines in reperfused renal tissue and increased serum superoxide dismutase levels to protect the kidney. The associated underlying mechanism may involve the inhibition of p38 kinase phosphorylation and c‑Jun N‑terminal kinase, anti‑lipid peroxidation, elimination of free radicals, and a reduction in the degree of apoptotic damage and oxidative stress injury induced by renal IRI. Therefore, Ru may be a suitable compound for the prevention and treatment of renal IRI.
Sigma-Aldrich
Anti-phospho-p38 MAPK (pTyr322) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-JNK1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-phospho-SAPK/JNK (pThr183) antibody produced in rabbit, affinity isolated antibody