MilliporeSigma
  • Efficient treatment of perfluorohexanoic acid by nanofiltration followed by electrochemical degradation of the NF concentrate.
Efficient treatment of perfluorohexanoic acid by nanofiltration followed by electrochemical degradation of the NF concentrate.
Water research (2017-02-06)
Álvaro Soriano, Daniel Gorri, Ane Urtiaga
ABSTRACT
The present study was aimed at the development of a strategy for removing and degrading perfluorohexanoic acid (PFHxA) from industrial process waters at concentrations in the range 60-200 mg L
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Undecafluorohexanoic acid, ≥97.0% (T)