MilliporeSigma
  • Putative Markers of Adulteration of Higher-Grade Olive Oil with Less Expensive Pomace Olive Oil Identified by Gas Chromatography Combined with Chemometrics.
Putative Markers of Adulteration of Higher-Grade Olive Oil with Less Expensive Pomace Olive Oil Identified by Gas Chromatography Combined with Chemometrics.
Journal of agricultural and food chemistry (2017-06-14)
Hazem Jabeur, Malika Drira, Ahmed Rebai, Mohamed Bouaziz
ABSTRACT
This work has been performed to ascertain that extra-virgin olive oil (EVOO) is free of adulteration. For this purpose, refined pomace olive oils (RPOOs) are commonly used for extra-virgin olive oil adulteration and repassed olive oils (ROOs) are used for lampante olive oil (LOO) fraudulent operation. Indeed, fatty acid ethyl esters could be used as a parameter for the detection of EVOO fraud with 2% RPOO. The addition of >10% RPOO to EVOO would be detected by the amount of erythrodiol, uvaol, waxes, and aliphatic alcohols. Moreover, the use of stigmasta-3,5-diene content proved to be effective in EVOO adulteration even at a low level (with 1% RPOO). For the detection of adulteration of LOO with >5% ROO, the sum of erythrodiol, uvaol, and the waxes and esters can be considered as good markers of purity. Using linear discriminant analysis can identify the most discriminant variable that allows a faster and cheaper evaluation of extra-virgin olive oil adulteration by measuring only these variables.
Sigma-Aldrich
Heptane, JIS special grade, ≥99.0%
Sigma-Aldrich
Dodecyl arachidate, ≥99%
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Cholesta-3,5-diene, ≥93% (HPLC)