MilliporeSigma
  • Different Bacterial Communities Involved in Peptide Decomposition between Normoxic and Hypoxic Coastal Waters.
Different Bacterial Communities Involved in Peptide Decomposition between Normoxic and Hypoxic Coastal Waters.
Frontiers in microbiology (2017-03-23)
Shuting Liu, Boris Wawrik, Zhanfei Liu
ABSTRACT
Proteins and peptides are key components of the labile dissolved organic matter pool in marine environments. Knowing which types of bacteria metabolize peptides can inform the factors that govern peptide decomposition and further carbon and nitrogen remineralization in marine environments. A
Sigma-Aldrich
21-Hydroxyprogesterone, ≥97% (HPLC)