MilliporeSigma
  • Synthesis and fluorescence spectroscopy of tris(pyrenyl)pnictogen compounds.
Synthesis and fluorescence spectroscopy of tris(pyrenyl)pnictogen compounds.
Dalton transactions (Cambridge, England : 2003) (2017-08-02)
Kira Behm, Jeremy B Essner, Charles L Barnes, Gary A Baker, Justin R Walensky
ABSTRACT
The synthesis, characterization, and fluorescence spectroscopy of E(C
Sigma-Aldrich
Quinine hemisulfate salt monohydrate, suitable for fluorescence, BioReagent, 99.0-101.0%