MilliporeSigma
  • Comparative in vitro and in vivo taste assessment of liquid praziquantel formulations.
Comparative in vitro and in vivo taste assessment of liquid praziquantel formulations.
International journal of pharmaceutics (2017-07-12)
Magdalena Münster, Abeer H A Mohamed-Ahmed, Laura I Immohr, Corinna Schoch, Carsten Schmidt, Catherine Tuleu, Jörg Breitkreutz
ABSTRACT
The taste of pharmaceuticals strongly affects the compliance of patients. This study investigated the applicability of the electronic tongue and rodent brief-access taste aversion (BATA) model for the bitter compound praziquantel (PZQ) and taste masked liquid formulations for PZQ. In a comparative study maltodextrin (MD) Kleptose
Sigma-Aldrich
Bis(2-ethylhexyl) phosphate, 97%
Sigma-Aldrich
Tetradodecylammonium bromide, ≥99.0% (AT)
Sigma-Aldrich
Methyltrioctylammonium chloride, ≥97.0% (AT)