MilliporeSigma
  • Evolutionary conserved mechanisms pervade structure and transcriptional modulation of allograft inflammatory factor-1 from sea anemone Anemonia viridis.
Evolutionary conserved mechanisms pervade structure and transcriptional modulation of allograft inflammatory factor-1 from sea anemone Anemonia viridis.
Fish & shellfish immunology (2017-06-06)
Angela Cuttitta, Maria Antonietta Ragusa, Salvatore Costa, Carmelo Bennici, Paolo Colombo, Salvatore Mazzola, Fabrizio Gianguzza, Aldo Nicosia
ABSTRACT
Gene family encoding allograft inflammatory factor-1 (AIF-1) is well conserved among organisms; however, there is limited knowledge in lower organisms. In this study, the first AIF-1 homologue from cnidarians was identified and characterised in the sea anemone Anemonia viridis. The full-length cDNA of AvAIF-1 was of 913 bp with a 5' -untranslated region (UTR) of 148 bp, a 3'-UTR of 315 and an open reading frame (ORF) of 450 bp encoding a polypeptide with149 amino acid residues and predicted molecular weight of about 17 kDa. The predicted protein possesses evolutionary conserved EF hand Ca
Sigma-Aldrich
Lipopolysaccharides from Pseudomonas aeruginosa 10, purified by trichloroacetic acid extraction
Sigma-Aldrich
Peptidoglycan from Bacillus subtilis