MilliporeSigma
  • Nitrated Fatty Acids Modulate the Physical Properties of Model Membranes and the Structure of Transmembrane Proteins.
Nitrated Fatty Acids Modulate the Physical Properties of Model Membranes and the Structure of Transmembrane Proteins.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-05-16)
Johannes Franz, Tristan Bereau, Stefanie Pannwitt, Veerappan Anbazhagan, Alexander Lehr, Udo Nubbemeyer, Ulrich Dietz, Mischa Bonn, Tobias Weidner, Dirk Schneider
ABSTRACT
Nitrated fatty acids (NO
Sigma-Aldrich
6-Dodecanoyl-N,N-dimethyl-2-naphthylamine, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
1,2-Dimyristoyl-d54-sn-glycero-3-phosphocholine, 98 atom % D, 97% (CP)
Sigma-Aldrich
Oleic acid, ≥99% (GC)