MilliporeSigma
  • Synthesis of novel indole derivatives as promising DNA-binding agents and evaluation of antitumor and antitopoisomerase I activities.
Synthesis of novel indole derivatives as promising DNA-binding agents and evaluation of antitumor and antitopoisomerase I activities.
European journal of medicinal chemistry (2017-05-23)
Elizabeth Almeida Lafayette, Sinara Mônica Vitalino de Almeida, Renata Virginia Cavalcanti Santos, Jamerson Ferreira de Oliveira, Cezar Augusto da Cruz Amorim, Rosali Maria Ferreira da Silva, Maira Galdino da Rocha Pitta, Ivan da Rocha Pitta, Ricardo Olimpio de Moura, Luiz Bezerra de Carvalho Júnior, Moacyr Jesus Barreto de Melo Rêgo, Maria do Carmo Alves de Lima
ABSTRACT
Molecules bearing indole nucleus present diverse biological properties such as antitumor and anti-inflammatory activities that can be associated both to DNA and protein interactions. This study focused on the synthesis of new indole derivatives with thiazolidines and imidazolidine rings condensed as side chains as well as the evaluation of their ability to interact with the DNA and antitumor and topoisomerase inhibition activities. All derivatives were successfully synthesized and their structures were elucidated by mass spectrometry (MS), infrared (IR), spectroscopy
Sigma-Aldrich
5-Bromoindole-3-carboxaldehyde, 98%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)