MilliporeSigma
  • Plant Phenotypic and Transcriptional Changes Induced by Volatiles from the Fungal Root Pathogen
Plant Phenotypic and Transcriptional Changes Induced by Volatiles from the Fungal Root Pathogen
Frontiers in plant science (2017-08-09)
Viviane Cordovez, Liesje Mommer, Kay Moisan, Dani Lucas-Barbosa, Ronald Pierik, Roland Mumm, Victor J Carrion, Jos M Raaijmakers
ABSTRACT
Beneficial soil microorganisms can affect plant growth and resistance by the production of volatile organic compounds (VOCs). Yet, little is known on how VOCs from soil-borne plant pathogens affect plant growth and resistance. Here we show that VOCs released from mycelium and sclerotia of the fungal root pathogen
Sigma-Aldrich
(±)-2-Methyl-1-butanol, ≥98.0% (GC)
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Supelco
2-Methyl-1-propanol, analytical standard