MilliporeSigma
  • Mutation I810N in the alpha3 isoform of Na+,K+-ATPase causes impairments in the sodium pump and hyperexcitability in the CNS.
Mutation I810N in the alpha3 isoform of Na+,K+-ATPase causes impairments in the sodium pump and hyperexcitability in the CNS.
Proceedings of the National Academy of Sciences of the United States of America (2009-08-12)
Steven J Clapcote, Steven Duffy, Gang Xie, Greer Kirshenbaum, Allison R Bechard, Vivien Rodacker Schack, Janne Petersen, Laleh Sinai, Bechara J Saab, Jason P Lerch, Berge A Minassian, Cameron A Ackerley, John G Sled, Miguel A Cortez, Jeffrey T Henderson, Bente Vilsen, John C Roder
ABSTRACT
In a mouse mutagenesis screen, we isolated a mutant, Myshkin (Myk), with autosomal dominant complex partial and secondarily generalized seizures, a greatly reduced threshold for hippocampal seizures in vitro, posttetanic hyperexcitability of the CA3-CA1 hippocampal pathway, and neuronal degeneration in the hippocampus. Positional cloning and functional analysis revealed that Myk/+ mice carry a mutation (I810N) which renders the normally expressed Na(+),K(+)-ATPase alpha3 isoform inactive. Total Na(+),K(+)-ATPase activity was reduced by 42% in Myk/+ brain. The epilepsy in Myk/+ mice and in vitro hyperexcitability could be prevented by delivery of additional copies of wild-type Na(+),K(+)-ATPase alpha3 by transgenesis, which also rescued Na(+),K(+)-ATPase activity. Our findings reveal the functional significance of the Na(+),K(+)-ATPase alpha3 isoform in the control of epileptiform activity and seizure behavior.