MilliporeSigma
  • Transgenic autoinhibition of p21-activated kinase exacerbates synaptic impairments and fronto-dependent behavioral deficits in an animal model of Alzheimer's disease.
Transgenic autoinhibition of p21-activated kinase exacerbates synaptic impairments and fronto-dependent behavioral deficits in an animal model of Alzheimer's disease.
Aging (2017-05-20)
Cyril Bories, Dany Arsenault, Myriam Lemire, Cyntia Tremblay, Yves De Koninck, Frédéric Calon
ABSTRACT
Defects in p21-activated kinase (PAK) lead to dendritic spine abnormalities and are sufficient to cause cognition impairment. The decrease in PAK in the brain of Alzheimer's disease (AD) patients is suspected to underlie synaptic and dendritic disturbances associated with its clinical expression, particularly with symptoms related to frontal cortex dysfunction. To investigate the role of PAK combined with Aβ and tau pathologies (3xTg-AD mice) in the frontal cortex, we generated a transgenic model of AD with a deficit in PAK activity (3xTg-AD-dnPAK mice). PAK inactivation had no effect on Aβ40 and Aβ42 levels, but increased the phosphorylation ratio of tau in detergent-insoluble protein fractions in the frontal cortex of 18-month-old heterozygous 3xTg-AD mice. Morphometric analyses of layer II/III pyramidal neurons in the frontal cortex showed that 3xTg-AD-dnPAK neurons exhibited significant dendritic attrition, lower spine density and longer spines compared to NonTg and 3xTg-AD mice. Finally, behavioral assessments revealed that 3xTg-AD-dnPAK mice exhibited pronounced anxious traits and disturbances in social behaviors, reminiscent of fronto-dependent symptoms observed in AD. Our results substantiate a critical role for PAK in the genesis of neuronal abnormalities in the frontal cortex underlying the emergence of psychiatric-like symptoms in AD.
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
Anti-Tau Antibody, a.a. 210-241, clone Tau-5, ascites fluid, clone Tau-5, Chemicon®
Sigma-Aldrich
Anti-GAD65 Antibody, from rabbit, purified by affinity chromatography