MilliporeSigma
  • Synthesis, characterisation and phase transition behaviour of temperature-responsive physically crosslinked poly (N-vinylcaprolactam) based polymers for biomedical applications.
Synthesis, characterisation and phase transition behaviour of temperature-responsive physically crosslinked poly (N-vinylcaprolactam) based polymers for biomedical applications.
Materials science & engineering. C, Materials for biological applications (2017-06-21)
Shane C Halligan, Maurice B Dalton, Kieran A Murray, Yixiao Dong, Wenxin Wang, John G Lyons, Luke M Geever
ABSTRACT
Poly (N-vinylcaprolactam) (PNVCL) is a polymer which offers superior characteristics for various potential medical device applications. In particular it offers unique thermoresponsive capabilities, which fulfils the material technology constraints required in targeted drug delivery applications. PNVCL phase transitions can be tailored in order to suit the requirements of current and next generation devices, by modifying the contents with regard to the material composition and aqueous polymer concentration. In this study, physically crosslinked Poly (N-vinylcaprolactam)-Vinyl acetate (PNVCL-VAc) copolymers were prepared by photopolymerisation. The structure of the polymers was established by Fourier transform infrared spectroscopy, nuclear magnetic resonance and gel permeation chromatography. The polymers were further characterised using differential scanning calorimetry and swelling studies. Determination of the LCST of the polymers in aqueous solution was achieved by employing four techniques; cloud point, UV-spectrometry, differential scanning calorimetry and rheometry. Sol-gel transition was established using tube inversion method and rheological analysis. This study was conducted to determine the characteristics of PNVCL with the addition of VAc, and to establish the effects on the phase transition. The PNVCL based polymers exhibited a decrease in the LCST as the composition of VAc increased. Sol-gel transition could be controlled by altering the monomeric feed ratio and polymer concentration in aqueous milieu. Importantly all copolymers (10wt% in solution) underwent gelation between 33.6 and 35.9°C, and based on this and the other materials properties recorded in this study, these novel copolymers have potential for use as injectable in situ forming drug delivery systems for targeted drug delivery.
Sigma-Aldrich
N-Vinylcaprolactam, 98%, stabilized