MilliporeSigma
  • The cleavage products of amyloid-beta precursor protein are sorted to distinct carrier vesicles that are independently transported within neurites.
The cleavage products of amyloid-beta precursor protein are sorted to distinct carrier vesicles that are independently transported within neurites.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2009-03-20)
Virgil Muresan, Nicholas H Varvel, Bruce T Lamb, Zoia Muresan
ABSTRACT
The amyloid-beta (Abeta) precursor protein (APP), a transmembrane protein that undergoes proteolytic cleavage into defined fragments, has been implicated in axonal transport. The proposed role of APP as a vesicle receptor for the microtubule motor kinesin-1 has relevance for the pathogenesis of Alzheimer's disease. Nevertheless, this function, which relies on the transport to the cell periphery of full-length APP rather than its cleavage fragments, remains controversial. Other proposed functions of APP, such as regulating transcription, neurogenesis, cell movement, or neurite growth also rely on APP's presence as a full-length protein at the cell surface, implying that APP cleavage occurs after its transport to the cell periphery. To test this hypothesis, we mapped the localization of various APP epitopes in neurons in culture and in the mouse brain. Surprisingly, epitopes from the N-terminal, C-terminal, and central (Abeta) domains of APP each showed a distinct distribution throughout the cell and rarely colocalized. Within neurites, these epitopes were localized to distinct transport vesicles that associated with different sets of microtubules and, occasionally, actin filaments. C-terminal APP fragments were preferentially transported into neurites as phosphorylated forms, entered the lamellipodium and filopodia of growth cones, and concentrated in regions of growth cone turning and advancement (unlike the N-terminal and Abeta fragments). We conclude that, under normal conditions, the proteolytic cleavage of APP primarily occurs before its sorting into axonal transport vesicles and the cleaved fragments segregate into separate vesicle populations that reach different destinations, and thus have different functions.
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-Amyloid Precursor Protein Antibody, CT, serum, Chemicon®
Sigma-Aldrich
Anti-Amyloid Precursor Protein Antibody, A4(695), clone 1.D5, clone 1.D5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Beta-Amyloid 1-42 Antibody, Chemicon®, from rabbit