MilliporeSigma
  • A new transformant selection system for the gray mold fungus Botrytis cinerea based on the expression of fenhexamid-insensitive ERG27 variants.
A new transformant selection system for the gray mold fungus Botrytis cinerea based on the expression of fenhexamid-insensitive ERG27 variants.
Fungal genetics and biology : FG & B (2017-02-12)
Kim Christopher Cohrs, Joachim Burbank, Julia Schumacher
ABSTRACT
The gray mold fungus Botrytis cinerea features a wide host range and causes severe economic losses, making it an important object for molecular research. Thus far, genetic modification of the fungus mainly is relied on two selection systems (nourseothricin and hygromycin), while other selection systems hold significant disadvantages. To broaden the spectrum of available molecular tools, a new selection system based on the cheap and widely used fungicide fenhexamid (hydroxyanilide group) was established. Fenhexamid specifically targets the 3-ketoreductase ERG27 from the ergosterol biosynthesis pathway. We generated a set of expression vectors suitable for deletion or expression of genes of interest (GOIs) in B. cinerea based on fenhexamid-insensitive ERG27 variants. Expression of BcERG27
Product Description
Supelco
Fenhexamid, PESTANAL®, analytical standard