MilliporeSigma
  • Core 1- and 3-derived O-glycans collectively maintain the colonic mucus barrier and protect against spontaneous colitis in mice.
Core 1- and 3-derived O-glycans collectively maintain the colonic mucus barrier and protect against spontaneous colitis in mice.
Mucosal immunology (2016-05-05)
K Bergstrom, J Fu, M E V Johansson, X Liu, N Gao, Q Wu, J Song, J M McDaniel, S McGee, W Chen, J Braun, G C Hansson, L Xia
ABSTRACT
Core 1- and 3-derived mucin-type O-glycans are primary components of the mucus layer in the colon. Reduced mucus thickness and impaired O-glycosylation are observed in human ulcerative colitis. However, how both types of O-glycans maintain mucus barrier function in the colon is unclear. We found that C1galt1 expression, which synthesizes core 1 O-glycans, was detected throughout the colon, whereas C3GnT, which controls core 3 O-glycan formation, was most highly expressed in the proximal colon. Consistent with this, mice lacking intestinal core 1-derived O-glycans (IEC C1galt1
Sigma-Aldrich
Tn Antigen, ≥99.0% (TLC)