MilliporeSigma
  • DHX9 suppresses RNA processing defects originating from the Alu invasion of the human genome.
DHX9 suppresses RNA processing defects originating from the Alu invasion of the human genome.
Nature (2017-03-30)
Tuğçe Aktaş, İbrahim Avşar Ilık, Daniel Maticzka, Vivek Bhardwaj, Cecilia Pessoa Rodrigues, Gerhard Mittler, Thomas Manke, Rolf Backofen, Asifa Akhtar
ABSTRACT
Transposable elements are viewed as 'selfish genetic elements', yet they contribute to gene regulation and genome evolution in diverse ways. More than half of the human genome consists of transposable elements. Alu elements belong to the short interspersed nuclear element (SINE) family of repetitive elements, and with over 1 million insertions they make up more than 10% of the human genome. Despite their abundance and the potential evolutionary advantages they confer, Alu elements can be mutagenic to the host as they can act as splice acceptors, inhibit translation of mRNAs and cause genomic instability. Alu elements are the main targets of the RNA-editing enzyme ADAR and the formation of Alu exons is suppressed by the nuclear ribonucleoprotein HNRNPC, but the broad effect of massive secondary structures formed by inverted-repeat Alu elements on RNA processing in the nucleus remains unknown. Here we show that DHX9, an abundant nuclear RNA helicase, binds specifically to inverted-repeat Alu elements that are transcribed as parts of genes. Loss of DHX9 leads to an increase in the number of circular-RNA-producing genes and amount of circular RNAs, translational repression of reporters containing inverted-repeat Alu elements, and transcriptional rewiring (the creation of mostly nonsensical novel connections between exons) of susceptible loci. Biochemical purifications of DHX9 identify the interferon-inducible isoform of ADAR (p150), but not the constitutively expressed ADAR isoform (p110), as an RNA-independent interaction partner. Co-depletion of ADAR and DHX9 augments the double-stranded RNA accumulation defects, leading to increased circular RNA production, revealing a functional link between these two enzymes. Our work uncovers an evolutionarily conserved function of DHX9. We propose that it acts as a nuclear RNA resolvase that neutralizes the immediate threat posed by transposon insertions and allows these elements to evolve as tools for the post-transcriptional regulation of gene expression.
Sigma-Aldrich
Anti-ZFR antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-MSL2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
MISSION® esiRNA, targeting human DHX9
Sigma-Aldrich
Monoclonal Anti-Splicing Factor SC-35 antibody produced in mouse, clone SC-35, ascites fluid
Sigma-Aldrich
Anti-ADAR antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution