MilliporeSigma
  • High-efficiency, environment-friendly electroluminescent polymers with stable high work function metal as a cathode: green- and yellow-emitting conjugated polyfluorene polyelectrolytes and their neutral precursors.
High-efficiency, environment-friendly electroluminescent polymers with stable high work function metal as a cathode: green- and yellow-emitting conjugated polyfluorene polyelectrolytes and their neutral precursors.
Journal of the American Chemical Society (2004-08-05)
Fei Huang, Lintao Hou, Hongbin Wu, Xiaohui Wang, Huilin Shen, Wei Cao, Wei Yang, Yong Cao
ABSTRACT
A series of aminoalkyl-substituted polyfluorene copolymers with benzothiadiazole (BTDZ) of different content were synthesized by Suzuki coupling reaction, and their quaternized ammonium polyelectrolyte derivatives were obtained through a postpolymerization treatment on the terminal amino groups. Copolymers are soluble in environmentally friendlier solvents, such as alcohols. It was found that the efficient energy transfer occurs by exciton trapping on the narrow band gap BTDZ site under UV illumination. Only 1% of BTDZ content is needed to completely quench a fluorene emission for both the neutral and the quaternized copolymers in the neat film. Absolute PL efficiencies of copolymer films were greatly enhanced as a result of the suppression of excimer formation. Light-emitting devices fabricated from these copolymers show high external quantum efficiencies over 3% and 1% for the neutral precursor and the quaternized copolymers, respectively, with high work function metals such as Al as a cathode. To the best of our knowledge, this is the first report on an electroluminescent polymer which bears the high EL efficiency, the electron-injection ability from high work function metals, and the solubility in environment-friendly solvents at the same time. These features make them a promising candidate for the next generation of light-emitting copolymers in PLED flat panel display application.
Sigma-Aldrich
PFN-DOF, Mw >10,000