MilliporeSigma
PEG-peptide conjugates.
Biomacromolecules (2014-04-12)
Ian W Hamley
ABSTRACT
The remarkable diversity of the self-assembly behavior of PEG-peptides is reviewed, including self-assemblies formed by PEG-peptides with β-sheet and α-helical (coiled-coil) peptide sequences. The modes of self-assembly in solution and in the solid state are discussed. Additionally, applications in bionanotechnology and synthetic materials science are summarized.
Sigma-Aldrich
Fmoc-N-amido-dPEG®8-acid, >95% (HPLC)
Sigma-Aldrich
Fmoc-N-amido-dPEG®2-acid, >95% (HPLC)
Sigma-Aldrich
Fmoc-N-amido-dPEG®36-acid, >95% (HPLC)
Sigma-Aldrich
Poly(ethylene glycol) α-hydroxy-ω-azido terminated, average Mn 5,000
Sigma-Aldrich
Poly(ethylene glycol) α-hydroxy-ω-azido terminated, average Mn 2,000