MilliporeSigma
  • New Mechanism for the Reduction of Vanadyl Acetylacetonate to Vanadium Acetylacetonate for Room Temperature Flow Batteries.
New Mechanism for the Reduction of Vanadyl Acetylacetonate to Vanadium Acetylacetonate for Room Temperature Flow Batteries.
ChemSusChem (2016-11-20)
Jack S Shamie, Caihong Liu, Leon L Shaw, Vincent L Sprenkle
ABSTRACT
In this study, a new mechanism for the reduction of vanadyl acetylacetonate, VO(acac)
Sigma-Aldrich
Vanadyl acetylacetonate, 98%