MilliporeSigma
  • Mitochondria-targeting "Nanoheater" for enhanced photothermal/chemo-therapy.
Mitochondria-targeting "Nanoheater" for enhanced photothermal/chemo-therapy.
Biomaterials (2016-12-13)
Si Chen, Qi Lei, Wen-Xiu Qiu, Li-Han Liu, Di-Wei Zheng, Jin-Xuan Fan, Lei Rong, Yun-Xia Sun, Xian-Zheng Zhang
ABSTRACT
In this work, mitochondria-targeting gold nanostar (AuNS) and anticarcinogen DOX were co-encapsulated in hyaluronic acid (HA) protective shell for tumor-targeting synergistic photothermal/chemo-therapy. Cationic peptide R
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Fmoc-D-Lys(Boc)-OH, 98%
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
HBTU, ≥98.0% (T)