MilliporeSigma
  • A Solution-Doped Polymer Semiconductor:Insulator Blend for Thermoelectrics.
A Solution-Doped Polymer Semiconductor:Insulator Blend for Thermoelectrics.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2017-01-21)
David Kiefer, Liyang Yu, Erik Fransson, Andrés Gómez, Daniel Primetzhofer, Aram Amassian, Mariano Campoy-Quiles, Christian Müller
ABSTRACT
Poly(ethylene oxide) is demonstrated to be a suitable matrix polymer for the solution-doped conjugated polymer poly(3-hexylthiophene). The polarity of the insulator combined with carefully chosen processing conditions permits the fabrication of tens of micrometer-thick films that feature a fine distribution of the F4TCNQ dopant:semiconductor complex. Changes in electrical conductivity from 0.1 to 0.3 S cm
Sigma-Aldrich
2,3,5,6-Tetrafluoro-7,7,8,8-tetracyanoquinodimethane, 97%