MilliporeSigma
  • Translational, rotational and vibrational relaxation dynamics of a solute molecule in a non-interacting solvent.
Translational, rotational and vibrational relaxation dynamics of a solute molecule in a non-interacting solvent.
Nature chemistry (2016-10-22)
Michael P Grubb, Philip M Coulter, Hugo J B Marroux, Balazs Hornung, Ryan S McMullen, Andrew J Orr-Ewing, Michael N R Ashfold
ABSTRACT
Spectroscopically observing the translational and rotational motion of solute molecules in liquid solutions is typically impeded by their interactions with the solvent, which conceal spectral detail through linewidth broadening. Here we show that unique insights into solute dynamics can be made with perfluorinated solvents, which interact weakly with solutes and provide a simplified liquid environment that helps to bridge the gap in our understanding of gas- and liquid-phase dynamics. Specifically, we show that in such solvents, the translational and rotational cooling of an energetic CN radical can be observed directly using ultrafast transient absorption spectroscopy. We observe that translational-energy dissipation within these liquids can be modelled through a series of classic collisions, whereas classically simulated rotational-energy dissipation is shown to be distinctly faster than experimentally measured. We also observe the onset of rotational hindering from nearby solvent molecules, which arises as the average rotational energy of the solute falls below the effective barrier to rotation induced by the solvent.
Sigma-Aldrich
Tetradecafluorohexane, 99%