MilliporeSigma
  • CD8+ T cells are crucial for the ability of congenic normal mice to reject highly immunogenic sarcomas induced in nude mice with 3-methylcholanthrene.
CD8+ T cells are crucial for the ability of congenic normal mice to reject highly immunogenic sarcomas induced in nude mice with 3-methylcholanthrene.
Clinical and experimental immunology (2000-08-10)
M Boesen, I M Svane, A M Engel, J Rygaard, A R Thomsen, O Werdelin
ABSTRACT
An attempt was made to identify the selection pressures put upon a growing tumour by CD8+ T cells. To this end tumours induced with 3-methylcholanthrene in T cell-deficient nude mice and in congenic T cell-competent nu/+ mice were transplanted to nu/+ recipients. The rejection rate of the sarcomas from nude mice was almost twice that of the sarcomas from nu/+ mice. Depletion of CD8+ T cells from nu/+ recipients prior to transplantation made them accept nude tumours that were consistently rejected by untreated nu/+ recipients. These findings suggest that a methylcholanthrene sarcoma during its growth in a T cell-competent host adapts to the T cell system through a selective elimination of highly immunogenic tumour cells that are susceptible to CD8+ T cell-mediated lysis.
Sigma-Aldrich
IgG from rat serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution