MilliporeSigma
  • Derivation, maintenance, and induction of the differentiation in vitro of equine embryonic stem cells.
Derivation, maintenance, and induction of the differentiation in vitro of equine embryonic stem cells.
Methods in molecular biology (Clifton, N.J.) (2006-07-19)
Shigeo Saito, Ken Sawai, Arika Minamihashi, Hideyo Ugai, Takehide Murata, Kazunari K Yokoyama
ABSTRACT
We describe here the isolation and maintenance of pluripotent embryonic stem (ES) cells from equine blastocysts that have been frozen and thawed. Equine ES cells appear to maintain a normal diploid karyotype in culture. These cells express markers that are characteristic of mouse ES cells, namely, alkaline phosphatase, stage-specific-embryonic antigen 1, STAT3, and Oct4. We also describe protocols for the induction of differentiation in vitro to neural precursor cells in the presence of basic fibroblast growth factor (bFGF), epidermal growth factor, and platelet-derived growth factor and to hematopoietic and endothelial cell lineages in the presence of bFGF, stem cell factor, and oncostatin M. Equine ES cells provide a powerful tool for gene targeting and the generation of transgenic clonal offspring.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Leukemia Inhibitory Factor human, LIF, recombinant, expressed in E. coli, 10 μg/ml, buffered aqueous solution (pH 7.4), suitable for cell culture
Sigma-Aldrich
Stem Cell Factor human, SCF, recombinant, expressed in E. coli, powder, suitable for cell culture