MilliporeSigma
  • Grb7 Protein Stability Modulated by Pin1 in Association with Cell Cycle Progression.
Grb7 Protein Stability Modulated by Pin1 in Association with Cell Cycle Progression.
PloS one (2016-09-23)
Yu-Ling Tai, Li-Hsuan Tung, Yu-Chi Lin, Pei-Jung Lu, Pei-Yu Chu, Ming-Yang Wang, Wei-Pang Huang, Ko-Chien Chen, Hsinyu Lee, Tang-Long Shen
ABSTRACT
Growth factor receptor bound protein-7 (Grb7) is a multi-domain adaptor protein that is co-opted by numerous tyrosine kinases involved in various cellular signaling and functions. The molecular mechanisms underlying the regulation of Grb7 remain unclear. Here, we revealed a novel negative post-translational regulation of Grb7 by the peptidyl-prolyl cis/trans isomerase, Pin1. Our data show that phosphorylation of Grb7 protein on the Ser194-Pro motif by c-Jun N-terminal kinase facilitates its binding with the WW domain of Pin1. Subsequently, Grb7 is degraded by the ubiquitin- and proteasome-dependent proteolytic pathway. Indeed, we found that Pin1 exerts its peptidyl-prolyl cis/trans isomerase activity in the modulation of Grb7 protein stability in regulation of cell cycle progression at the G2-M phase. This study illustrates a novel regulatory mechanism in modulating Grb7-mediated signaling, which may take part in pathophysiological consequences.
Sigma-Aldrich
Monoclonal Anti-BrdU antibody produced in mouse, clone BU-33, ascites fluid, Immunohistology Grade
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-phospho-p38α(Thr180/Tyr182) Antibody, clone 8.78.8, rabbit monoclonal, culture supernatant, clone 8.78.8, from rabbit
Sigma-Aldrich
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, clone MPM-2, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-Glutathione-S-Transferase (GST) antibody produced in mouse, clone GST-2, ascites fluid
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®