MilliporeSigma
  • Exome sequencing of hepatocellular carcinomas identifies new mutational signatures and potential therapeutic targets.
Exome sequencing of hepatocellular carcinomas identifies new mutational signatures and potential therapeutic targets.
Nature genetics (2015-03-31)
Kornelius Schulze, Sandrine Imbeaud, Eric Letouzé, Ludmil B Alexandrov, Julien Calderaro, Sandra Rebouissou, Gabrielle Couchy, Clément Meiller, Jayendra Shinde, Frederic Soysouvanh, Anna-Line Calatayud, Roser Pinyol, Laura Pelletier, Charles Balabaud, Alexis Laurent, Jean-Frederic Blanc, Vincenzo Mazzaferro, Fabien Calvo, Augusto Villanueva, Jean-Charles Nault, Paulette Bioulac-Sage, Michael R Stratton, Josep M Llovet, Jessica Zucman-Rossi
ABSTRACT
Genomic analyses promise to improve tumor characterization to optimize personalized treatment for patients with hepatocellular carcinoma (HCC). Exome sequencing analysis of 243 liver tumors identified mutational signatures associated with specific risk factors, mainly combined alcohol and tobacco consumption and exposure to aflatoxin B1. We identified 161 putative driver genes associated with 11 recurrently altered pathways. Associations of mutations defined 3 groups of genes related to risk factors and centered on CTNNB1 (alcohol), TP53 (hepatitis B virus, HBV) and AXIN1. Analyses according to tumor stage progression identified TERT promoter mutation as an early event, whereas FGF3, FGF4, FGF19 or CCND1 amplification and TP53 and CDKN2A alterations appeared at more advanced stages in aggressive tumors. In 28% of the tumors, we identified genetic alterations potentially targetable by US Food and Drug Administration (FDA)-approved drugs. In conclusion, we identified risk factor-specific mutational signatures and defined the extensive landscape of altered genes and pathways in HCC, which will be useful to design clinical trials for targeted therapy.
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-NQO1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution