MilliporeSigma
  • Raising the N-aryl fluoride content in unsymmetrical diaryliminoacenaphthylenes as a route to highly active nickel(ii) catalysts in ethylene polymerization.
Raising the N-aryl fluoride content in unsymmetrical diaryliminoacenaphthylenes as a route to highly active nickel(ii) catalysts in ethylene polymerization.
Dalton transactions (Cambridge, England : 2003) (2016-11-03)
Xinxin Wang, Linlin Fan, Yichun Yuan, Shizhen Du, Yang Sun, Gregory A Solan, Cun-Yue Guo, Wen-Hua Sun
ABSTRACT
Five examples of selectively fluorinated unsymmetrical diiminoacenaphthylenes, 1-[2,6-{(4-FC
Sigma-Aldrich
Ethylaluminum sesquichloride, 97%