MilliporeSigma
Identification of myoglobin in human smooth muscle.
The Journal of biological chemistry (1998-08-29)
Y Qiu, L Sutton, A F Riggs
ABSTRACT
Myoglobin (Mb) has been believed to be absent generally from mammalian smooth muscle tissue. Examination of human rectal, uterine, bladder, colon, small intestine, arterial, and venous smooth muscle by immunohistochemical techniques shows that each of these tissues is immunopositive for both smooth muscle myosin and human Mb. Mb-specific primers were used for the polymerase chain reaction to generate cDNA from smooth muscle tissues. Southern hybridization with a Mb-specific probe gave a very strong signal with the cDNA from rectum, weaker signals from small intestine and uterus, a faint signal from colon, and no signal from bladder tissue. High performance liquid chromatography analysis coupled with sequence determination has shown that contaminating heme-binding serum albumin as well as hemoglobin in extracts of smooth muscle seriously compromise any heme-based or spectrophotometric assay of Mb. Combined affinity and size exclusion chromatography, however, provide the necessary resolution. The cDNA-derived amino acid sequence of human smooth muscle Mb was found to be identical to that of Mb from striated muscle.
Sigma-Aldrich
Bovine Serum Albumin, pH <5.0, powder