MilliporeSigma
Inhibition of acetylcholinesterase by the anticancer prodrug CPT-11.
Chemico-biological interactions (2005-11-01)
Janice L Hyatt, Lyudmila Tsurkan, Christopher L Morton, Kyoung J P Yoon, Michal Harel, Boris Brumshtein, Israel Silman, Joel L Sussman, Randy M Wadkins, Philip M Potter
ABSTRACT
CPT-11 (irinotecan, 7-ethyl-10-[4-(1-piperidino)-1-piperidino]carbonyloxycamptothecin) is an anticancer prodrug that has been approved for the treatment of colon cancer. It is a member of the camptothecin class of drugs and activation to the active metabolite SN-38, is mediated by carboxylesterases (CE). SN-38 is a potent topoisomerase I poison and is highly effective at killing human tumor cells, with IC50 values in the low nM range. However, upon high dose administration of CPT-11 to cancer patients, a cholinergic syndrome is observed, that can be rapidly ameliorated by atropine. This suggests a direct interaction of the drug or its metabolites with acetylcholinesterase (AChE). Kinetic studies indicated that CPT-11 was primarily responsible for AChE inhibition with the 4-piperidinopiperidine moiety, the major determinant in the loss of enzyme activity. Structural analogs of 4-piperidinopiperidine however, did not inhibit AChE, including a benzyl piperazine derivate of CPT-11. These results suggest that novel anticancer drugs could be synthesized that do not inhibit AChE, or alternatively, that novel AChE inhibitors could be designed based around the camptothecin scaffold.
Sigma-Aldrich
4-Piperidinopiperidine, 97%