MilliporeSigma
  • Protein tyrosine phosphatases: functional inferences from mouse models and human diseases.
Protein tyrosine phosphatases: functional inferences from mouse models and human diseases.
The FEBS journal (2008-02-27)
Wiljan J A J Hendriks, Ari Elson, Sheila Harroch, Andrew W Stoker
ABSTRACT
Some 40-odd genes in mammals encode phosphotyrosine-specific, 'classical' protein tyrosine phosphatases. The generation of animal model systems and the study of various human disease states have begun to elucidate the important and diverse roles of protein tyrosine phosphatases in cellular signalling pathways, development and disease. Here, we provide an overview of those findings from mice and men, and indicate several novel approaches that are now being exploited to further our knowledge of this fascinating enzyme family.
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Bacterial Glycerol Stock, Phosphatases
Sigma-Aldrich
MISSION® shRNA Mouse Gene Family Set, Lentiviral Particles, Phosphatases
Sigma-Aldrich
MISSION® shRNA Mouse Gene Family Set, Bacterial Glycerol Stock, Phosphatases
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, DNA, Phosphatases
Sigma-Aldrich
MISSION® shRNA Mouse Gene Family Set, DNA, Phosphatases
Sigma-Aldrich
MISSION® shRNA Human Gene Family Set, Lentiviral Particles, Phosphatases