MilliporeSigma
  • Microvesicles provide a mechanism for intercellular communication by embryonic stem cells during embryo implantation.
Microvesicles provide a mechanism for intercellular communication by embryonic stem cells during embryo implantation.
Nature communications (2016-06-16)
Laura M Desrochers, François Bordeleau, Cynthia A Reinhart-King, Richard A Cerione, Marc A Antonyak
ABSTRACT
Communication between the inner cell mass (ICM) and the trophoblast layer of the blastocyst is known to occur, but its functional consequences on early developmental events is unclear. Here we demonstrate that embryonic stem (ES) cells derived from the ICM generate and shed microvesicles (MVs), a major class of extracellular vesicles (EVs), which influence trophoblast behaviour during the implantation process. The MV cargo proteins laminin and fibronectin interact with integrins along the surfaces of the trophoblasts, triggering the activation of two signalling kinases, JNK and FAK, and stimulating trophoblast migration. We further show that injecting MVs isolated from ES cells into blastocysts results in an increase in their implantation efficiency. Thus, these findings highlight a unique mechanism by which ES cells communicate with trophoblasts within the blastocyst to increase their ability to migrate into the uterus, thereby promoting one of the earliest and most important steps during pregnancy.
Sigma-Aldrich
Monoclonal Anti-Fibronectin, Cellular antibody produced in mouse, clone FN-3E2, ascites fluid
Sigma-Aldrich
Anti-Rac1 Antibody, clone 23A8, clone 23A8, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-LAMA5 antibody produced in mouse, clone 2F7, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid