MilliporeSigma
  • Myoscape controls cardiac calcium cycling and contractility via regulation of L-type calcium channel surface expression.
Myoscape controls cardiac calcium cycling and contractility via regulation of L-type calcium channel surface expression.
Nature communications (2016-04-29)
Matthias Eden, Benjamin Meder, Mirko Völkers, Montatip Poomvanicha, Katrin Domes, M Branchereau, P Marck, Rainer Will, Alexander Bernt, Ashraf Rangrez, Matthias Busch, Martin Hrabě de Angelis, Christophe Heymes, Wolfgang Rottbauer, Patrick Most, Franz Hofmann, Norbert Frey
ABSTRACT
Calcium signalling plays a critical role in the pathogenesis of heart failure. Here we describe a cardiac protein named Myoscape/FAM40B/STRIP2, which directly interacts with the L-type calcium channel. Knockdown of Myoscape in cardiomyocytes decreases calcium transients associated with smaller Ca(2+) amplitudes and a lower diastolic Ca(2+) content. Likewise, L-type calcium channel currents are significantly diminished on Myoscape ablation, and downregulation of Myoscape significantly reduces contractility of cardiomyocytes. Conversely, overexpression of Myoscape increases global Ca(2+) transients and enhances L-type Ca(2+) channel currents, and is sufficient to restore decreased currents in failing cardiomyocytes. In vivo, both Myoscape-depleted morphant zebrafish and Myoscape knockout (KO) mice display impairment of cardiac function progressing to advanced heart failure. Mechanistically, Myoscape-deficient mice show reduced L-type Ca(2+)currents, cell capacity and calcium current densities as a result of diminished LTCC surface expression. Finally, Myoscape expression is reduced in hearts from patients suffering of terminal heart failure, implying a role in human disease.
Product Description
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Anti-STRIP2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution