MilliporeSigma
The α5 subunit containing GABAA receptors contribute to chronic pain.
Pain (2015-11-07)
Mariana Bravo-Hernández, José A Corleto, Paulino Barragán-Iglesias, Ricardo González-Ramírez, Jorge B Pineda-Farias, Ricardo Felix, Nigel A Calcutt, Rodolfo Delgado-Lezama, Martin Marsala, Vinicio Granados-Soto
ABSTRACT
It has been recently proposed that α5-subunit containing GABAA receptors (α5-GABAA receptors) that mediate tonic inhibition might be involved in pain. The purpose of this study was to investigate the contribution of α5-GABAA receptors in the loss of GABAergic inhibition and in formalin-induced, complete Freund's adjuvant (CFA)-induced and L5 and L6 spinal nerve ligation-induced long-lasting hypersensitivity. Formalin or CFA injection and L5 and L6 spinal nerve ligation produced long-lasting allodynia and hyperalgesia. Moreover, formalin injection impaired the rate-dependent depression of the Hofmann reflex. Peripheral and intrathecal pretreatment or post-treatment with the α5-GABAA receptor antagonist, L-655,708 (0.15-15 nmol), prevented and reversed, respectively, these long-lasting behaviors. Formalin injection increased α5-GABAA receptor mRNA expression in the spinal cord and dorsal root ganglia (DRG) mainly at 3 days. The α5-GABAA receptors were localized in the dorsal spinal cord and DRG colabeling with NeuN, CGRP, and IB4 which suggests their presence in peptidergic and nonpeptidergic neurons. These receptors were found mainly in small and medium sized neurons. Formalin injection enhanced α5-GABAA receptor fluorescence intensity in spinal cord and DRG at 3 and 6 days. Intrathecal administration of L-655,708 (15 nmol) prevented and reversed formalin-induced impairment of rate-dependent depression. These results suggest that α5-GABAA receptors play a role in the loss of GABAergic inhibition and contribute to long-lasting secondary allodynia and hyperalgesia.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP)−Cy3 antibody, Mouse monoclonal, clone G-A-5, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NeuN Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-GABRA5 (ab2) antibody produced in rabbit, affinity isolated antibody