MilliporeSigma
  • Organolithium-induced alkylative ring opening of aziridines: synthesis of unsaturated amino alcohols and ethers.
Organolithium-induced alkylative ring opening of aziridines: synthesis of unsaturated amino alcohols and ethers.
The Journal of organic chemistry (2006-10-27)
David M Hodgson, Bogdan Stefane, Timothy J Miles, Jason Witherington
ABSTRACT
Organolithium-induced alkylative ring opening of N-sulfonyl-protected aziridinyl ethers is described. The reactions were efficiently carried out with a variety of organolithiums, providing a promising new strategy to unsaturated amino alcohols and ethers. Cis- and trans-1,4-dimethoxybut-2-ene-derived aziridines were prepared, and their propensity to undergo organolithium- induced alkylative desymmetrization is detailed. Use of a single enantiomer of the latter aziridine provides a route to enantiopure unsaturated amino ethers.
Sigma-Aldrich
Isopropyllithium solution, 0.7 M in pentane