MilliporeSigma
  • Metabolic evolution and (13) C flux analysis of a succinate dehydrogenase deficient strain of Yarrowia lipolytica.
Metabolic evolution and (13) C flux analysis of a succinate dehydrogenase deficient strain of Yarrowia lipolytica.
Biotechnology and bioengineering (2016-05-18)
Tigran V Yuzbashev, Pavel Yu Bondarenko, Tatiana I Sobolevskaya, Evgeniya Yu Yuzbasheva, Ivan A Laptev, Vadim V Kachala, Alexander S Fedorov, Tatiana V Vybornaya, Anna S Larina, Sergey P Sineoky
ABSTRACT
Bio-based succinic acid production can redirect industrial chemistry processes from using limited hydrocarbons to renewable carbohydrates. A fermentation process that does not require pH-titrating agents will be advantageous to the industry. Previously, a Yarrowia lipolytica strain that was defective for succinate dehydrogenase was constructed and was found to accumulate up to 17.5 g L(-1) of succinic acid when grown on glycerol without buffering. Here, a derivative mutant was isolated that produced 40.5 g L(-1) of succinic acid in 36 h with a yield of 0.32 g g(-1) glycerol. A combination approach of induced mutagenesis and metabolic evolution allowed isolation of another derivative that could utilize glucose efficiently and accumulated 50.2 g L(-1) succinic acid in 54 h with a yield of 0.43 g g(-1) . The parent strain of these isolated mutants was used for [1,6-(13) C2 ]glucose assimilation analysis. At least 35% glucose was estimated to be utilized through the pentose phosphate pathway, while ≥84% succinic acid was formed through the oxidative branch of the tricarboxylic acid cycle. Biotechnol. Bioeng. 2016;113: 2425-2432. © 2016 Wiley Periodicals, Inc.