MilliporeSigma
  • Axially chiral guanidine as highly active and enantioselective catalyst for electrophilic amination of unsymmetrically substituted 1,3-dicarbonyl compounds.
Axially chiral guanidine as highly active and enantioselective catalyst for electrophilic amination of unsymmetrically substituted 1,3-dicarbonyl compounds.
Journal of the American Chemical Society (2006-12-15)
Masahiro Terada, Megumi Nakano, Hitoshi Ube
ABSTRACT
A newly designed axially chiral guanidine is found to function as an effective platform for asymmetric induction at the alpha-carbon of unsymmetrically substituted 1,3-dicarbonyl compounds. Highly efficient and enantioselective electrophilic amination of various 1,3-dicarbonyl compounds with azodicarboxylate was successfully achieved using the present chiral guanidine catalyst, which provides efficient access to the construction of nitrogen-substituted quaternary stereocenters in an optically active form.
Sigma-Aldrich
Di-tert-butyl azodicarboxylate, 98%