MilliporeSigma
  • Indium(I) iodide promoted cleavage of diphenyl diselenide and disulfide and subsequent palladium(0)-catalyzed condensation with vinylic bromides. A simple one-pot synthesis of vinylic selenides and sulfides.
Indium(I) iodide promoted cleavage of diphenyl diselenide and disulfide and subsequent palladium(0)-catalyzed condensation with vinylic bromides. A simple one-pot synthesis of vinylic selenides and sulfides.
The Journal of organic chemistry (2006-01-04)
Brindaban C Ranu, Kalicharan Chattopadhyay, Subhash Banerjee
ABSTRACT
[reaction: see text] Diphenyl diselenide (and disulfide) undergo facile reaction with indium(I) iodide and the corresponding intermediate complex condenses in situ with a variety of substituted vinyl bromides in the presence of a catalytic amount of tetrakis(triphenylphosphine)palladium(0) [Pd(PPh3)4] in THF at room temperature to produce vinylic selenides and sulfides in good yields. The conversion of (E)-vinyl bromides is remarkably stereoselective giving (E)-vinyl selenides (and sulpfides) whereas the stereoselectivity in reaction of (Z)-vinyl bromides is not very good.
Sigma-Aldrich
Indium(I) iodide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.