MilliporeSigma
  • Biofunctionalized zinc oxide field effect transistors for selective sensing of riboflavin with current modulation.
Biofunctionalized zinc oxide field effect transistors for selective sensing of riboflavin with current modulation.
Sensors (Basel, Switzerland) (2011-12-14)
Joshua A Hagen, Sang N Kim, Burhan Bayraktaroglu, Kevin Leedy, Jorge L Chávez, Nancy Kelley-Loughnane, Rajesh R Naik, Morley O Stone
ABSTRACT
Zinc oxide field effect transistors (ZnO-FET), covalently functionalized with single stranded DNA aptamers, provide a highly selective platform for label-free small molecule sensing. The nanostructured surface morphology of ZnO provides high sensitivity and room temperature deposition allows for a wide array of substrate types. Herein we demonstrate the selective detection of riboflavin down to the pM level in aqueous solution using the negative electrical current response of the ZnO-FET by covalently attaching a riboflavin binding aptamer to the surface. The response of the biofunctionalized ZnO-FET was tuned by attaching a redox tag (ferrocene) to the 3' terminus of the aptamer, resulting in positive current modulation upon exposure to riboflavin down to pM levels.
Sigma-Aldrich
2-Quinoxalinecarboxylic acid, 97%