MilliporeSigma
  • Self-inactivating lentivirus vector for safe and efficient in vivo gene delivery.
Self-inactivating lentivirus vector for safe and efficient in vivo gene delivery.
Journal of virology (1998-11-13)
R Zufferey, T Dull, R J Mandel, A Bukovsky, D Quiroz, L Naldini, D Trono
ABSTRACT
In vivo transduction of nondividing cells by human immunodeficiency virus type 1 (HIV-1)-based vectors results in transgene expression that is stable over several months. However, the use of HIV-1 vectors raises concerns about their safety. Here we describe a self-inactivating HIV-1 vector with a 400-nucleotide deletion in the 3' long terminal repeat (LTR). The deletion, which includes the TATA box, abolished the LTR promoter activity but did not affect vector titers or transgene expression in vitro. The self-inactivating vector transduced neurons in vivo as efficiently as a vector with full-length LTRs. The inactivation design achieved in this work improves significantly the biosafety of HIV-derived vectors, as it reduces the likelihood that replication-competent retroviruses will originate in the vector producer and target cells, and hampers recombination with wild-type HIV in an infected host. Moreover, it improves the potential performance of the vector by removing LTR sequences previously associated with transcriptional interference and suppression in vivo and by allowing the construction of more-stringent tissue-specific or regulatable vectors.
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Transduction Particles, Targets no known mammalian genes
Sigma-Aldrich
MISSION® pLKO.1-puro TurboGFP shRNA Control Plasmid DNA, shRNA sequence targeting tGFP
Sigma-Aldrich
MISSION® pLKO.1-puro eGFP shRNA Control Plasmid DNA, shRNA sequence targeting eGFP
Sigma-Aldrich
MISSION® pLKO.1-puro eGFP shRNA Control Transduction Particles, shRNA sequence targeting eGFP