MilliporeSigma
  • ATP induces NO production in hippocampal neurons by P2X(7) receptor activation independent of glutamate signaling.
ATP induces NO production in hippocampal neurons by P2X(7) receptor activation independent of glutamate signaling.
PloS one (2013-03-09)
Juan Francisco Codocedo, Juan Alejandro Godoy, Maria Ines Poblete, Nibaldo C Inestrosa, Juan Pablo Huidobro-Toro
ABSTRACT
To assess the putative role of adenosine triphosphate (ATP) upon nitric oxide (NO) production in the hippocampus, we used as a model both rat hippocampal slices and isolated hippocampal neurons in culture, lacking glial cells. In hippocampal slices, additions of exogenous ATP or 2'(3')-O-(4-Benzoylbenzoyl) ATP (Bz-ATP) elicited concentration-dependent NO production, which increased linearly within the first 15 min and plateaued thereafter; agonist EC50 values were 50 and 15 µM, respectively. The NO increase evoked by ATP was antagonized in a concentration-dependent manner by Coomassie brilliant blue G (BBG) or by N(ω)-propyl-L-arginine, suggesting the involvement of P2X7Rs and neuronal NOS, respectively. The ATP induced NO production was independent of N-methyl-D-aspartic acid (NMDA) receptor activity as effects were not alleviated by DL-2-Amino-5-phosphonopentanoic acid (APV), but antagonized by BBG. In sum, exogenous ATP elicited NO production in hippocampal neurons independently of NMDA receptor activity.
Sigma-Aldrich
2′(3′)-O-(4-Benzoylbenzoyl)adenosine 5′-triphosphate triethylammonium salt, ≥93%