MilliporeSigma
  • Synthesis and insecticidal activity of benzoheterocyclic analogues of N'-benzoyl-N-(tert-butyl)benzohydrazide: Part 1. Design of benzoheterocyclic analogues.
Synthesis and insecticidal activity of benzoheterocyclic analogues of N'-benzoyl-N-(tert-butyl)benzohydrazide: Part 1. Design of benzoheterocyclic analogues.
Pest management science (2003-02-01)
Yoshihiro Sawada, Toshiaki Yanai, Harumi Nakagawa, Yoshihisa Tsukamoto, Shinji Yokoi, Mikio Yanagi, Tetsuya Toya, Hiroyasu Sugizaki, Yasuhito Kato, Hidetoshi Shirakura, Tetsuo Watanabe, Yoshimi Yajima, Seiichirou Kodama, Akio Masui
ABSTRACT
The N'-benzoyl group of N-tert-butyl-N'-benzoyl-3,5-dimethylbenzohydrazide (1) was converted to a series of benzoheterocyclecarbonyl groups in order to investigate the potential usefulness of superimposing a hydrazine insecticide on 20-hydroxyecdysone. A series of analogues with benzodioxole, benzodioxane, benzodioxapine, indole, benzoxazole, benzoxazine or benzothiazole instead of the phenyl group of (1) were synthesized and tested for their insecticidal activity against the common cutworm (Spodoptera litura F). N-tert-Butyl-N'-(3,5-dimethylbenzoyl)-1,3-benzodioxole-5-carbohydrazide and N-tert-butyl-N'-(3,5-dimethylbenzoyl)-2,3-dihydro-1,4-benzodioxine-6-carbohydrazide showed high insecticidal activities, superior to that of (1) and equal to that of the commercial insecticide tebufenozide (RH-5992).
Sigma-Aldrich
Benzothiazole-6-carboxylic acid, 96%