MilliporeSigma
Heterocyclic bibenzimidazole derivatives as topoisomerase I inhibitors.
Bioorganic & medicinal chemistry letters (2000-04-27)
S Jin, J S Kim, S P Sim, A Liu, D S Pilch, L F Liu, E J LaVoie
ABSTRACT
A series of 2'-heterocyclic derivatives of 5-phenyl-2,5'-1H-bibenzimidazoles were evaluated for topoisomerase I poisoning activity and cytotoxicity. Topo I poisoning activity was associated with 2'-derivatives that possessed a hydrogen atom capable of hydrogen bond formation, suggesting that the interatomic distances between such hydrogen atoms and the heteroatoms on the adjacent benzimidazole influence activity.
Sigma-Aldrich
Indole-5-carboxaldehyde, 98%