MilliporeSigma
  • Expression of human apolipoprotein A-I in transgenic mice results in reduced plasma levels of murine apolipoprotein A-I and the appearance of two new high density lipoprotein size subclasses.
Expression of human apolipoprotein A-I in transgenic mice results in reduced plasma levels of murine apolipoprotein A-I and the appearance of two new high density lipoprotein size subclasses.
Proceedings of the National Academy of Sciences of the United States of America (1991-01-15)
E M Rubin, B Y Ishida, S M Clift, R M Krauss
ABSTRACT
In Western societies high density lipoprotein (HDL) levels correlate inversely with the risk for coronary heart disease. The primary protein component of both human and mouse HDL is apolipoprotein A-I (apoAI), which comprises greater than 70% of HDL protein and 30% of HDL mass. Human HDLs include particles of several distinct size subpopulations, whereas HDLs from inbred C57BL/6 mice contain a single population of particles. To study the regulation of apoAI expression and its role in HDL assembly, we created transgenic C57BL/6 mice containing the human apoAI gene. Two independent lines of transgenic mice with approximately twice the normal plasma levels of total apoAI were studied. The level of mouse apoAI is reduced greater than 4-fold in both transgenic lines, comprising only 4% of total plasma apoAI levels in one transgenic line and 13% in the other. We demonstrate that the mechanism responsible for the decrease in mouse apoAI is posttranscriptional. Parallel to the replacement of mouse with human apoAI, the single HDL species normally present in the plasma of C57BL/6 is replaced by two HDL subclasses similar in size to human HDL2b and HDL3a. The changes in murine apolipoprotein levels and HDL subclass size are inherited by all transgenic offspring of the two founder animals. These results suggest a dominant role of apoAI in determining the HDL particle size distribution and a mechanism involving expression of human apoAI transgenes that alters the plasma levels of mouse apoAI.
Sigma-Aldrich
SILuLite APOA1 Apolipoprotein A-1 human, recombinant, expressed in HEK 293 cells, MS Protein Standard