MilliporeSigma
  • Pharmacokinetics of oseltamivir: an oral antiviral for the treatment and prophylaxis of influenza in diverse populations.
Pharmacokinetics of oseltamivir: an oral antiviral for the treatment and prophylaxis of influenza in diverse populations.
The Journal of antimicrobial chemotherapy (2010-03-20)
Brian E Davies
ABSTRACT
Influenza is a transmissible viral pathogen that continues to cause substantial morbidity and mortality. Oseltamivir is an orally administered antiviral medication that selectively inhibits the influenza neuraminidase enzymes that are essential for viral replication. Treatment of infected children > or =1 year and adults of all ages may decrease the severity and duration of the symptoms of infection, while prophylactic dosing can prevent their onset. Oseltamivir is ingested in the form of a prodrug (oseltamivir phosphate) that is rapidly converted by hepatic esterases into the active metabolite, oseltamivir carboxylate. Oseltamivir carboxylate has high bioavailability and penetrates sites of infection at concentrations that are sufficient to inhibit viral replication. The pharmacokinetics of oseltamivir and oseltamivir carboxylate are dose proportional after repeated doses of up to 500 mg twice daily. This predictable profile means that oseltamivir is suitable for use in diverse patient populations, which may include young children and elderly patients, various ethnic groups and those with renal or hepatic impairment. As the potential for drug interactions is low, oseltamivir is also suitable for use in patients with co-morbid conditions who are likely to be receiving concomitant medications.
Sigma-Aldrich
Oseltamivir phosphate, ≥98% (HPLC)