MilliporeSigma
  • A mechanistic investigation on the utilization of lactose as a protective agent for multi-unit pellet systems.
A mechanistic investigation on the utilization of lactose as a protective agent for multi-unit pellet systems.
Pharmaceutical development and technology (2014-12-19)
Wun Chyi Chin, Lai Wah Chan, Paul Wan Sia Heng
ABSTRACT
The effect of lactose particle size on the extent of pellet coat damage was investigated. The extent of pellet coat damage increased linearly with lactose median particle size. It was observed that coated pellets compressed with coarser lactose grades had larger and deeper surface indentations. The surfaces of the pellets compressed with coarser lactose grades were also found to be significantly rougher. Micronized lactose was capable of protecting pellet coats from damage brought about by the presence of coarser lactose particles. The findings suggested a protective effect that micronized lactose conferred to pellet coats was not only through dimensional delimitations but also by higher interparticulate friction and longer particle rearrangement phase. As a result, the pellet volume fraction in the system was reduced. The extent of pellet coat damage was found to escalate when the pellet volume fraction in such system increased beyond a critical value of 0.39.
Sigma-Aldrich
Sodium hydroxyacetate, AldrichCPR